missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ Closures for Dropper Bottles: Bulk Pack
GREENER_CHOICE

Catalog No. 0339066F
Change view
Click to view available options
Color:
Blue
Green
Natural
Red
White
Yellow
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
0339066F Blue
0339066A Natural
0339066B White
0339066C Yellow
0339066D Green
0339066E Red
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 0339066F Supplier Thermo Scientific™ Supplier No. 3127600060
Only null left

Order these closures, which are available in multiple colors to help identify reagents, to complete your dropper bottle system.

Order Thermo Scientific™ Nalgene™ Closures for Dropper Bottles to complete the Nalgene Dropper Bottle System. Closures fit dropper bottles Cat. Nos. 312750 and 31275, and are available in multiple colors to help identify reagents.

Features:
  • Polypropylene
  • Available in various colors

Ordering Information:

Dropper Bottle Closures are bulk packed 2000 per case. Order Nalgene controlled-drop dispensing tip (Cat. Nos. 312758, 312759) and dropper bottles (Cat. Nos. 312750, 312751) separately.

Specifications

Color Blue
Material Polypropylene
No. per Case 2000
No. per Pack 2000
Neck Finish 15-415
Certifications/Compliance Manufactured in compliance with ISO 13485:2003 quality requirements. Dropper Bottle Closures meet the requirements of 21CFR177.1520 for food and beverage use, USP Class VI.
Closure Included Yes
Product Type Dropping Bottle Screw Cap
For Use With (Equipment) Dropper Bottles 312750 and 312751.
Closure Material Polypropylene