missing translation for 'onlineSavingsMsg'
Learn More

Nabumetone, MilliporeSigma™ Supelco™

Catalog No. 111014187
Change view
Click to view available options
Quantity:
5 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014187 5 g
111014186 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111014187 Supplier Merck Emd Millipore Supplier No. N61425G
Featured Industries: Pharmaceutical (small molecule)

Specifications

CAS 42924-53-8
Physical Form Neat
Linear Formula C15H16O2
MDL Number MFCD00079518
Synonym 4-(6-Methoxy-2-naphthyl)-2-butanone
Type Analytical Standard
Quantity 5 g
Formula Weight 228.29
Grade Analytical Standard