missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals N6-Benzyladenosin

Catalog No. AC226420050
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC226420050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC226420050 Supplier Thermo Scientific Chemicals Supplier No. 226420050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

 

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Molecular Weight (g/mol) 357.36
Product Type N6-Benzyladenosine
Quantity 5 g
Solubility Information Solubility aq. soln.: insoluble. Other solubilities: soluble in ethanol, aq. hcl/ethanol, aq. naoh/etha, nol

RUO – Research Use Only