missing translation for 'onlineSavingsMsg'
Learn More

N6-Benzyladenine 99.0+%, TCI America™

Catalog No. B10885G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B10885G 5 g
B108825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B10885G Supplier TCI America Supplier No. B10885G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1214-39-7
Molecular Formula C12H11N5
Molecular Weight (g/mol) 225.26
MDL Number MFCD00005572
InChI Key NWBJYWHLCVSVIJ-UHFFFAOYSA-N
Synonym 6-benzylaminopurine, benzyladenine, n6-benzyladenine, n-benzyladenine, n-benzyl-9h-purin-6-amine, 6-benzyladenine, 6-benzylamino purine, cytokinin b, adenine, n-benzyl, 6-bap
PubChem CID 62389
ChEBI CHEBI:29022
IUPAC Name N-benzyl-7H-purin-6-amine
SMILES C(NC1=C2NC=NC2=NC=N1)C1=CC=CC=C1

Specifications

Melting Point 234°C
Color White
Quantity 5 g
Formula Weight 225.26
Percent Purity ≥99.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N6-Benzyladenine
EINECSNumber (5)-1137
RTECSNumber AU6252200
TSCA Yes