missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N6-Benzoyl-5'-O-(4,4'-dimethoxytrityl)-2'-deoxyadenosine 99.0+%, TCI America™
Specifications
Specifications
| Melting Point | 94°C |
| Color | White |
| Quantity | 1 g |
| Synonym | n6-benzoyl-5'-o-4,4'-dimethoxytrityl-2'-deoxyadenosine, dmbta, bz-dmt-da, n 6-benzoyl-5'-o-4,4'-dimethoxytrityl-2'-deoxyadenosine, adenosine, n-benzoyl-5'-o-bis 4-methoxyphenyl phenylmethyl-2'-deoxy, n-9-2r,4s,5r-5-bis 4-methoxyphenyl phenyl methoxy methyl-4-hydroxyoxolan-2-yl purin-6-yl benzamide, ksc621a9d, n-benzoyl-5'-o-bis 4-methoxyphenyl phenylmethyl-2'-deoxyadenosine, 5'-ortho-dmt-n6-bz-2'-deoxyadenosine |
| Formula Weight | 657.73 |
| Percent Purity | ≥99.0% (HPLC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | N6-Benzoyl-5′-O-(4,4′-dimethoxytrityl)-2′-deoxyadenosine |
Safety and Handling
| TSCA | Yes |