missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N4-Benzoylcytidine 98.0+%, TCI America™
$55.84 - $173.26
Chemical Identifiers
| CAS | 13089-48-0 |
|---|---|
| Molecular Formula | C16H17N3O6 |
| Molecular Weight (g/mol) | 347.33 |
| MDL Number | MFCD00010572,MFCD01457402 |
| InChI Key | BNXBRFDWSPXODM-UHFFFAOYNA-N |
| Synonym | n6-benzoylcytidine, n-1-2r,4r,5r-3,4-dihydroxy-5-hydroxymethyl oxolan-2-yl-2-oxopyrimidin-4-yl benzamide |
| PubChem CID | 133611947 |
| IUPAC Name | N-{1-[3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-2-oxo-1,2-dihydropyrimidin-4-yl}benzamide |
| SMILES | OCC1OC(C(O)C1O)N1C=CC(NC(=O)C2=CC=CC=C2)=NC1=O |
Chemical Identifiers
| 13089-48-0 | |
| 347.33 | |
| BNXBRFDWSPXODM-UHFFFAOYNA-N | |
| 133611947 | |
| OCC1OC(C(O)C1O)N1C=CC(NC(=O)C2=CC=CC=C2)=NC1=O |
| C16H17N3O6 | |
| MFCD00010572,MFCD01457402 | |
| n6-benzoylcytidine, n-1-2r,4r,5r-3,4-dihydroxy-5-hydroxymethyl oxolan-2-yl-2-oxopyrimidin-4-yl benzamide | |
| N-{1-[3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-2-oxo-1,2-dihydropyrimidin-4-yl}benzamide |
Specifications
| 13089-48-0 | |
| White | |
| MFCD00010572,MFCD01457402 | |
| n6-benzoylcytidine, n-1-2r,4r,5r-3,4-dihydroxy-5-hydroxymethyl oxolan-2-yl-2-oxopyrimidin-4-yl benzamide | |
| OCC1OC(C(O)C1O)N1C=CC(NC(=O)C2=CC=CC=C2)=NC1=O | |
| 347.33 | |
| 347.33 | |
| Crystalline Powder |
| 240°C | |
| C16H17N3O6 | |
| 1 g | |
| BNXBRFDWSPXODM-UHFFFAOYNA-N | |
| N-{1-[3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-2-oxo-1,2-dihydropyrimidin-4-yl}benzamide | |
| 133611947 | |
| ≥98.0% (HPLC,T) | |
| N4-Benzoylcytidine |
Safety and Handling
TSCA : No