missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N4-Benzoylcytidine 98.0+%, TCI America™
Supplier: TCI America B30945G
Specifications
| N4-Benzoylcytidine | |
| 240°C | |
| C16H17N3O6 | |
| 5 g | |
| BNXBRFDWSPXODM-UHFFFAOYNA-N | |
| N-{1-[3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-2-oxo-1,2-dihydropyrimidin-4-yl}benzamide | |
| 133611947 | |
| ≥98.0% (HPLC,T) |
| 13089-48-0 | |
| White | |
| MFCD00010572,MFCD01457402 | |
| n6-benzoylcytidine, n-1-2r,4r,5r-3,4-dihydroxy-5-hydroxymethyl oxolan-2-yl-2-oxopyrimidin-4-yl benzamide | |
| OCC1OC(C(O)C1O)N1C=CC(NC(=O)C2=CC=CC=C2)=NC1=O | |
| 347.33 | |
| 347.33 | |
| Crystalline Powder |
Chemical Identifiers
| 13089-48-0 | |
| 347.33 | |
| BNXBRFDWSPXODM-UHFFFAOYNA-N | |
| 133611947 | |
| OCC1OC(C(O)C1O)N1C=CC(NC(=O)C2=CC=CC=C2)=NC1=O |
| C16H17N3O6 | |
| MFCD00010572,MFCD01457402 | |
| n6-benzoylcytidine, n-1-2r,4r,5r-3,4-dihydroxy-5-hydroxymethyl oxolan-2-yl-2-oxopyrimidin-4-yl benzamide | |
| N-{1-[3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-2-oxo-1,2-dihydropyrimidin-4-yl}benzamide |
Safety and Handling
TSCA : No