accessibility menu, dialog, popup

UserName

CAS RN 13089-48-0

HOOHOHONNOHNO

CAS RN 13089-48-0

IUPAC Name: N-{1-[3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-2-oxo-1,2-dihydropyrimidin-4-yl}benzamide
Synonyms: cytidine, N-benzoyl-
Molecular Weight (g/mol): 347.33
Molecular Formula: C16H17N3O6
InChi Key: BNXBRFDWSPXODM-UHFFFAOYNA-N
SMILES: OCC1OC(C(O)C1O)N1C=CC(NC(=O)C2=CC=CC=C2)=NC1=O
Percent Purity 
  • (2)
  • (3)
Physical Form 
  • (5)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (5)
  • (1)
  • (1)
  • (1)
  • (2)