missing translation for 'onlineSavingsMsg'
Learn More

N4-Acetylcytosine 98.0+%, TCI America™

Catalog No. A20895G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A20895G 5 g
A208925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A20895G Supplier TCI America Supplier No. A20895G

Chemical Identifiers

CAS 14631-20-0
Molecular Formula C6H7N3O2
Molecular Weight (g/mol) 153.14
MDL Number MFCD00134466
InChI Key IJCKBIINTQEGLY-UHFFFAOYSA-N
Synonym n4-acetylcytosine, n-acetylcytosine, n-2-oxo-1,2-dihydropyrimidin-4-yl acetamide, acetamide, n-1,2-dihydro-2-oxo-4-pyrimidinyl, n 4-acetylcytosine, n-2-oxo-1h-pyrimidin-6-yl acetamide, n-2-hydroxypyrimidin-4-yl acetamide, n-2-oxo-3h-pyrimidin-4-yl acetamide, 4-acetylamino-1,2-dihydro-2-pyrimidone
PubChem CID 99309
IUPAC Name N-(2-oxo-2,3-dihydropyrimidin-4-yl)acetamide
SMILES CC(=O)NC1=CC=NC(=O)N1

Specifications

Melting Point 325°C
Color White
Quantity 5 g
Formula Weight 153.14
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material N4-Acetylcytosine