missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N4-Acetylcytosine, 98%, Thermo Scientific™
Spécifications
Spécifications
| Nom ou substance chimique | N4-Acetylcytosine |
| Spectre infrarouge | Authentic |
| Fourchette de pourcentages d’essai | 97.5% min. (HPLC) |
| Quantité | 25g |
| Poids de formule | 153.14 |
| Pourcentage de pureté | 98% |
Sécurité et manipulation
| Hazard Category | GHS Signal Word: Warning |
| Hazard Statement | GHS H Statement Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | GHS P Statement IF ON SKIN: Wash with plenty of soap and water. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses,if present and easy to do. Continue rinsing. IF INHALED: Remove to fresh air and kee |
RUO – Research Use Only