missing translation for 'onlineSavingsMsg'
Learn More

N1-(4-Fluorobenzyl)-3-nitro-1,4-phenylenediamine 97.0+%, TCI America™

Catalog No. F10825G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F10825G 5 g
F10821G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F10825G Supplier TCI America Supplier No. F10825G
Only null left

Chemical Identifiers

CAS 150812-21-8
Molecular Formula C13H12FN3O2
Molecular Weight (g/mol) 261.256
MDL Number MFCD12828054
InChI Key XTDZJOIEYRRRGJ-UHFFFAOYSA-N
Synonym n1-4-fluorobenzyl-3-nitrobenzene-1,4-diamine, 1-n-4-fluorophenyl methyl-3-nitrobenzene-1,4-diamine, 2-amino-5-4-fluorobenzyl amino-1-nitrobenzene, fluorophenylmethylnitrobenzenediamine, n1-4-fluorophenyl methyl-3-nitrobenzene-1,4-diamine, 4-4-fluorobenzylamino-2-nitroaniline, n-4-fluorobenzyl-3-nitro-p-phenylenediamine, n1-4-fluorobenzyl-3-nitro-1,4-phenylenediamine, n∼4∼-4-fluorophenyl methyl-2-nitrobenzene-1,4-diamine, n4-4-fluorophenyl methyl-2-nitro-benzene-1,4-diamine
PubChem CID 20096587
IUPAC Name 4-N-[(4-fluorophenyl)methyl]-2-nitrobenzene-1,4-diamine
SMILES C1=CC(=CC=C1CNC2=CC(=C(C=C2)N)[N+](=O)[O-])F

Specifications

Melting Point 112°C
Quantity 5 g
Formula Weight 261.26
Percent Purity ≥97.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N1-(4-Fluorobenzyl)-3-nitro-1,4-phenylenediamine