missing translation for 'onlineSavingsMsg'
Learn More

N^w-Nitro-L-arginine, 98%

Catalog No. AAH6206114
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6206114 25 g
AAH6206106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6206114 Supplier Thermo Scientific Chemicals Supplier No. H6206114
Get Up to $1,800 in Chemicals with Purchase
View all promotions

missing translation for 'validMainframeMsg'

CAS: 2149-70-4 | C6H13N5O4 | 219.201 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2149-70-4
Molecular Formula C6H13N5O4
Molecular Weight (g/mol) 219.201
MDL Number MFCD00007033
InChI Key MRAUNPAHJZDYCK-BYPYZUCNSA-N
Synonym h-arg no2-oh, ng-nitro-l-arginine, l-nna, nitroarginine, n-nitro-l-arginine, nomega-nitro-l-arginine, n g-nitro-l-arginine, ng-no2-l-arg, n-omega-nitro-l-arginine, n g-nitroarginine
PubChem CID 440005
ChEBI CHEBI:27960
SMILES C(CC(C(=O)O)N)CN=C(N)N[N+](=O)[O-]

Specifications

Melting Point 257°C
Beilstein 1728914
Quantity 25 g
Formula Weight 219.2
Percent Purity 98%
Chemical Name or Material Nomega-Nitro-L-arginine
EINECSNumber 218-418-9
RTECSNumber RM2982100
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only