missing translation for 'onlineSavingsMsg'
Learn More

N-Tris(hydroxymethyl)methyl-2-aminoethanesulfonic Acid 98.0+%, TCI America™

Catalog No. T068325G
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T068325G 25 g
T0683250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T068325G Supplier TCI America Supplier No. T068325G

[Good's buffer component for biological research]

Chemical Identifiers

CAS 7365-44-8
Molecular Formula C6H15NO6S
Molecular Weight (g/mol) 229.247
MDL Number MFCD00007532
InChI Key JOCBASBOOFNAJA-UHFFFAOYSA-N
Synonym tes, n-tris hydroxymethyl methyl-2-aminoethanesulfonic acid, tes buffer, tes free acid, ethanesulfonic acid, 2-2-hydroxy-1,1-bis hydroxymethyl ethyl amino, 2-1,3-dihydroxy-2-hydroxymethyl propan-2-yl amino ethanesulfonic acid, 2-2-hydroxy-1,1-dihydroxymethyl-ethylamino-ethanesulfonic acid, tris hydroxymethyl methyl-2-aminomethane sulfonic acid, 2-tris hydroxymethyl methylamino ethane-1-sulphonic acid, n-tri hydroxymethyl methyltaurine
PubChem CID 81831
ChEBI CHEBI:44356
IUPAC Name 2-[[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino]ethanesulfonic acid
SMILES C(CS(=O)(=O)O)NC(CO)(CO)CO

Specifications

Melting Point 216°C
Color White
Quantity 25 g
Formula Weight 229.25
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material N-Tris(hydroxymethyl)methyl-2-aminoethanesulfonic Acid [Good′s buffer component for biological