missing translation for 'onlineSavingsMsg'
Learn More

N-Thionylaniline, 96%

Catalog No. AAH5945114
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5945114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH5945114 Supplier Thermo Scientific Chemicals Supplier No. H5945114
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1122-83-4 | C6H5NOS | 139.172 g/mol

It is a versatile synthetic intermediate and an active pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a versatile synthetic intermediate and an active pharmaceutical intermediate.

Solubility
Partly miscible in water.

Notes
Store at room temperature. Keep away from oxidizing agents and heats.

Chemical Identifiers

CAS 1122-83-4
Molecular Formula C6H5NOS
Molecular Weight (g/mol) 139.172
MDL Number MFCD00002084
InChI Key FIOJWGRGPONADF-UHFFFAOYSA-N
Synonym n-thionylaniline, n-sulfinylaniline, benzenamine, n-sulfinyl, thionylaniline, phenylthionylamine, phenylsulfinylamine, sulfinylphenylamine, aniline, n-sulfinyl, thionyl imide, phenyl, sulfurous imide, phenyl
PubChem CID 70739
IUPAC Name (sulfinylamino)benzene
SMILES C1=CC=C(C=C1)N=S=O

Specifications

Color Brown
Density 1.236 g/mL
Boiling Point 200°C
Flash Point 84°C (183°F)
Refractive Index 1.6275
Quantity 25 g
Beilstein 507682
Solubility Information Partly miscible in water.
Formula Weight 139.18
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material N-Thionylaniline
Show More Show Less
Hazard Category H227
Hazard Statement GHS H Statement
H334-H335-H227-H315-H319-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause respiratory irritation.
Combustible liquid.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
Precautionary Statement P210-P280-P370+P378q-P501c
EINECSNumber 214-362-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only