missing translation for 'onlineSavingsMsg'
Learn More

N-(tert-Butyl)hydroxylamine hydrochloride, 98%

Catalog No. AC186910010
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC186910010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC186910010 Supplier Thermo Scientific Chemicals Supplier No. 186910010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 57497-39-9 | C4H11NO·HCl | 125.6 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 57497-39-9
Molecular Formula C4H11NO·HCl
Molecular Weight (g/mol) 125.6
MDL Number MFCD00012598
InChI Key DCSATTBHEMKGIP-UHFFFAOYSA-N
Synonym n-tert-butyl hydroxylamine hydrochloride, n-tert-butyl hydroxylamine hcl, n-tert-butylhydroxylamine hydrochloride, n-1,1-dimethylethyl hydroxylamine hydrochloride, 2-propanamine, n-hydroxy-2-methyl-, hydrochloride, 2-methylprop-2-ylhydroxylamine, chloride, soyheadhjuup@, acmc-1b1z0, t-butylhydroxylamine hydrochloride
PubChem CID 2777896
IUPAC Name N-tert-butylhydroxylamine;hydrochloride
SMILES CC(C)(C)NO.Cl

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 176.0°C to 180.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (Argentometry)
Packaging Glass bottle
Linear Formula (CH3)3CNHOH·HCl
Quantity 1 g
Fieser 12,95; 04,191; 05,47; 06,64; 08,49; 10,49; 14,52; 17,50
Formula Weight 125.6
Percent Purity 98%
Physical Form Powder
Chemical Name or Material N-(tert-Butyl)hydroxylamine hydrochloride
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 260-771-6

RUO – Research Use Only