missing translation for 'onlineSavingsMsg'
Learn More

N-tert-Butyl-alpha-phenylnitrone 98.0+%, TCI America™

Catalog No. B17015G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B17015G 5 g
B17011G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B17015G Supplier TCI America Supplier No. B17015G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 3376-24-7
Molecular Formula C11H15NO
Molecular Weight (g/mol) 177.247
MDL Number MFCD00008799
InChI Key IYSYLWYGCWTJSG-FMIVXFBMSA-N
Synonym e-n-benzylidene-2-methylpropan-2-amine oxide
PubChem CID 10313352
IUPAC Name N-tert-butyl-1-phenylmethanimine oxide
SMILES CC(C)(C)[N+](=CC1=CC=CC=C1)[O-]

Specifications

Melting Point 74°C
Quantity 5 g
Formula Weight 177.25
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-tert-Butyl-alpha-phenylnitrone