missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-tert-Butyl-alpha-(4-pyridyl-1-oxide)nitrone 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 185°C |
| Color | White-Yellow |
| Quantity | 5 g |
| Formula Weight | 194.23 |
| Percent Purity | ≥98.0% (HPLC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | N-tert-Butyl-alpha-(4-pyridyl-1-oxide)nitrone |
Safety and Handling
| TSCA | No |
| Recommended Storage | Refrigerator |