missing translation for 'onlineSavingsMsg'
Learn More

N-(tert-Butoxycarbonyl)-trans-4-(p-toluenesulfonyloxy)-L-proline Methyl Ester 98.0+%, TCI America™

Catalog No. B5247200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B5247200MG 200 mg
B52471G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B5247200MG Supplier TCI America Supplier No. B5247200MG
Only null left

Chemical Identifiers

CAS 88043-21-4
Molecular Formula C18H25NO7S
Molecular Weight (g/mol) 399.458
MDL Number MFCD02179383
InChI Key UKVKNGDXVREPDE-HIFRSBDPSA-N
Synonym boc-trans-4-tosyloxy-l-proline methyl ester, n-boc-trans-4-tosyloxy-l-proline methyl ester, 4r-4-tso-n-boc-pro-ome, n-boc-trans-4-p-tosyloxy-l-proline methyl ester, 2s,4r-1-tert-butyl 2-methyl 4-tosyloxy pyrrolidine-1,2-dicarboxylate, 2s,4r-4-4-methylphenyl sulfonyl oxy-1,2-pyrrolidinedicarboxylic acid 1-1,1-dimethylethyl 2-methyl ester, 1-tert-butyl 2-methyl 2s,4r-4-4-methylbenzenesulfonyl oxy pyrrolidine-1,2-dicarboxylate, ksc659e9j, boc-trans-4-tosyloxy-l-prolinemethylester
PubChem CID 13425772
IUPAC Name 1-O-tert-butyl 2-O-methyl (2S,4R)-4-(4-methylphenyl)sulfonyloxypyrrolidine-1,2-dicarboxylate
SMILES CC1=CC=C(C=C1)S(=O)(=O)OC2CC(N(C2)C(=O)OC(C)(C)C)C(=O)OC

Specifications

Melting Point 79°C
Color White-Yellow
Quantity 200 mg
Formula Weight 399.46
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material N-(tert-Butoxycarbonyl)-trans-4-(p-toluenesulfonyloxy)-L-proline Methyl Ester
TSCA No
Recommended Storage Refrigerator