missing translation for 'onlineSavingsMsg'
Learn More

N-(tert-Butoxycarbonyl)-L-tryptophan 98.0+%, TCI America™

Catalog No. B163925G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B163925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B163925G Supplier TCI America Supplier No. B163925G

Chemical Identifiers

CAS 13139-14-5
Molecular Formula C16H20N2O4
Molecular Weight (g/mol) 304.35
MDL Number MFCD000655
InChI Key NFVNYBJCJGKVQK-ZDUSSCGKSA-N
Synonym boc-trp-oh, n-boc-l-tryptophan, boc-l-tryptophan, n-tert-butoxy carbonyl-l-tryptophan, boc-l-trp-oh, boc-l-tryptophane, s-2-tert-butoxycarbonyl amino-3-1h-indol-3-yl propanoic acid, l-tryptophan, n-1,1-dimethylethoxy carbonyl, boc-tryptophan, n-tert-butoxycarbonyl-l-tryptophan
PubChem CID 83169
IUPAC Name (2S)-2-{[(tert-butoxy)carbonyl]amino}-3-(1H-indol-3-yl)propanoic acid
SMILES CC(C)(C)OC(=O)N[C@@H](CC1=CNC2=CC=CC=C12)C(O)=O

Specifications

Melting Point 133°C
Quantity 25 g
Formula Weight 304.35
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material N-(tert-Butoxycarbonyl)-L-tryptophan