missing translation for 'onlineSavingsMsg'
Learn More

N-(tert-Butoxycarbonyl)-L-methionine 98.0+%, TCI America™

Catalog No. B163610G
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B163610G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B163610G Supplier TCI America Supplier No. B163610G

Chemical Identifiers

CAS 2488-15-5
Molecular Formula C10H19NO4S
Molecular Weight (g/mol) 249.33
MDL Number MFCD00065586
InChI Key IMUSLIHRIYOHEV-ZETCQYMHSA-N
Synonym boc-l-methionine, boc-met-oh, n-boc-l-methionine, n-tert-butoxycarbonyl-l-methionine, tert-butoxycarbonyl-l-methionine, s-2-tert-butoxycarbonyl amino-4-methylthio butanoic acid, n-alpha-t-butyloxycarbonyl-l-methionine, methionine, n-1,1-dimethylethoxy carbonyl, l-methionine, n-1,1-dimethylethoxy carbonyl
PubChem CID 89857
IUPAC Name (2S)-2-{[(tert-butoxy)carbonyl]amino}-4-(methylsulfanyl)butanoic acid
SMILES CSCC[C@H](NC(=O)OC(C)(C)C)C(O)=O

Specifications

Melting Point 50°C
Color White
Quantity 10 g
Formula Weight 249.33
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-(tert-Butoxycarbonyl)-L-methionine