missing translation for 'onlineSavingsMsg'
Learn More

N-(tert-Butoxycarbonyl)-L-alanine 98.0+%, TCI America™

Catalog No. B11845G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B11845G 5 g
B118425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B11845G Supplier TCI America Supplier No. B11845G

N-(tert-Butoxycarbonyl)-L-alanine, >-98.0% (HPLC,T), C8H15NO4, CAS Number-15761-38-3, boc-ala-oh, boc-l-alanine, tert-butoxycarbonyl-l-alanine, boc-l-ala-oh, tert-butoxycarbonyl alanine, boc-ala, n-tert-butoxycarbonyl-l-alanine, n-t-boc-l-alanine, n-boc-l-alanine, n-alpha-t-boc-l-alanine, 25g, White

N-(tert-Butoxycarbonyl)-L-alanine, >-98.0% (HPLC,T), C8H15NO4, CAS Number-15761-38-3, boc-ala-oh, boc-l-alanine, tert-butoxycarbonyl-l-alanine, boc-l-ala-oh, tert-butoxycarbonyl alanine, boc-ala, n-tert-butoxycarbonyl-l-alanine, n-t-boc-l-alanine, n-boc-l-alanine, n-alpha-t-boc-l-alanine, 25g, White

Chemical Identifiers

CAS 15761-38-3
Molecular Formula C8H15NO4
Molecular Weight (g/mol) 189.21
MDL Number MFCD00037225
InChI Key QVHJQCGUWFKTSE-YFKPBYRVSA-N
Synonym boc-ala-oh, n-tert-butoxycarbonyl-l-alanine, boc-l-alanine, n-boc-l-alanine, boc-l-ala-oh, n-t-boc-l-alanine, boc-ala, tert-butoxycarbonyl-l-alanine, tert-butoxycarbonyl alanine, n-alpha-t-boc-l-alanine
PubChem CID 85082
IUPAC Name (2S)-2-{[(tert-butoxy)carbonyl]amino}propanoic acid
SMILES C[C@H](NC(=O)OC(C)(C)C)C(O)=O

Specifications

Melting Point 83°C
Color White
Quantity 5 g
Formula Weight 189.21
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-(tert-Butoxycarbonyl)-L-alanine