missing translation for 'onlineSavingsMsg'
Learn More

N-(tert-Butoxycarbonyl)iminodiacetic Acid 98.0+%, TCI America™

Catalog No. B43685G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B43685G 5 g
B436825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B43685G Supplier TCI America Supplier No. B43685G
Only null left

Chemical Identifiers

CAS 56074-20-5
Molecular Formula C9H15NO6
Molecular Weight (g/mol) 233.22
MDL Number MFCD00800351
InChI Key QPGIJQUTGABQLQ-UHFFFAOYSA-N
Synonym n-boc-iminodiacetic acid, boc-dl-ida-oh, 2,2'-tert-butoxycarbonyl azanediyl diacetic acid, tert-butoxycarbonyl carboxymethyl amino acetic acid, n-tert-butoxycarbonyl iminodiacetic acid, glycine, n-carboxymethyl-n-1,1-dimethylethoxy carbonyl, 2-carboxymethyl-2-methylpropan-2-yl oxycarbonyl amino acetic acid, tert-butoxycarbonylimino bisacetic acid, n-t-butyloxycarbonyl iminodiacetic acid, n-tert-butoxycarbonyl-iminodiacetic acid
PubChem CID 2755971
IUPAC Name 2-[carboxymethyl-[(2-methylpropan-2-yl)oxycarbonyl]amino]acetic acid
SMILES CC(C)(C)OC(=O)N(CC(=O)O)CC(=O)O

Specifications

Melting Point 126°C
Color White
Quantity 5 g
Formula Weight 233.22
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-(tert-Butoxycarbonyl)iminodiacetic Acid
TSCA No
Recommended Storage Refrigerator