missing translation for 'onlineSavingsMsg'
Learn More

N-(tert-Butoxycarbonyl)glycine 98.0+%, TCI America™

Catalog No. B11855G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B11855G 5 g
B118525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B11855G Supplier TCI America Supplier No. B11855G

Chemical Identifiers

CAS 4530-20-5
Molecular Formula C7H13NO4
Molecular Weight (g/mol) 175.18
MDL Number MFCD00002690
InChI Key VRPJIFMKZZEXLR-UHFFFAOYSA-N
Synonym boc-glycine, boc-gly-oh, n-tert-butoxycarbonyl glycine, n-boc-glycine, n-boc-l-glycine, tert-butoxycarbonylglycine, t-butoxycarbonylglycine, 2-tert-butoxycarbonyl amino acetic acid, glycine, n-1,1-dimethylethoxy carbonyl, tert-butyloxycarbonylglycine
PubChem CID 78288
IUPAC Name 2-{[(tert-butoxy)carbonyl]amino}acetic acid
SMILES CC(C)(C)OC(=O)NCC(O)=O

Specifications

Melting Point 89°C
Color White
Quantity 5 g
Formula Weight 175.18
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material N-(tert-Butoxycarbonyl)glycine