missing translation for 'onlineSavingsMsg'
Learn More

N-(tert-Butoxycarbonyl)-D-tryptophan 98.0+%, TCI America™

Catalog No. B22595G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B22595G 5 g
B22591G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B22595G Supplier TCI America Supplier No. B22595G

Chemical Identifiers

CAS 5241-64-5
Molecular Formula C16H20N2O4
Molecular Weight (g/mol) 304.35
MDL Number MFCD00037944
InChI Key NFVNYBJCJGKVQK-CYBMUJFWSA-N
Synonym boc-d-trp-oh, n-boc-d-tryptophan, n-tert-butoxy carbonyl-d-tryptophan, n-tert-butoxycarbonyl-d-tryptophan, boc-d-tryptophan, boc-d-trp, nalpha-boc-d-tryptophan, boc-d-tryptophane, nalpha-boc-d-tryptophane, tert-butoxycarbonyl
PubChem CID 111050
IUPAC Name (2R)-2-{[(tert-butoxy)carbonyl]amino}-3-(1H-indol-3-yl)propanoic acid
SMILES CC(C)(C)OC(=O)N[C@H](CC1=CNC2=CC=CC=C12)C(O)=O

Specifications

Color White
Quantity 5 g
Formula Weight 304.35
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material N-(tert-Butoxycarbonyl)-D-tryptophan