missing translation for 'onlineSavingsMsg'
Learn More

N-(tert-Butoxycarbonyl)-D-threonine 98.0+%, TCI America™

Catalog No. B29905G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B29905G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B29905G Supplier TCI America Supplier No. B29905G

Chemical Identifiers

CAS 55674-67-4
Molecular Formula C9H17NO5
Molecular Weight (g/mol) 219.24
MDL Number MFCD00037807
InChI Key LLHOYOCAAURYRL-NTSWFWBYSA-N
Synonym boc-d-thr-oh, boc-d-threonine, n-tert-butoxycarbonyl-d-threonine, n-boc-d-threonine, 2r,3s-2-tert-butoxycarbonyl amino-3-hydroxybutanoic acid, n-alpha-tert-butoxycarbonyl-d-threonine, d-threonine, n-1,1-dimethylethoxy carbonyl, pubchem14945, boc-d-thr
PubChem CID 6992541
IUPAC Name (2R,3S)-2-{[(tert-butoxy)carbonyl]amino}-3-hydroxybutanoic acid
SMILES C[C@H](O)[C@@H](NC(=O)OC(C)(C)C)C(O)=O

Specifications

Melting Point 81°C
Color White
Quantity 5 g
Formula Weight 219.24
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-(tert-Butoxycarbonyl)-D-threonine