missing translation for 'onlineSavingsMsg'
Learn More

N-(tert-Butoxycarbonyl)-D-serine Methyl Ester 98.0+%, TCI America™

Catalog No. B48595G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B48595G 5 g
B48591G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B48595G Supplier TCI America Supplier No. B48595G

Chemical Identifiers

CAS 95715-85-8
Molecular Formula C9H17NO5
Molecular Weight (g/mol) 219.237
MDL Number MFCD00270516
InChI Key SANNKFASHWONFD-ZCFIWIBFSA-N
Synonym boc-d-ser-ome, boc-d-serine methyl ester, n-boc-d-serine methyl ester, n-boc-d-serine methylester, n-tert-butoxycarbonyl-d-serine methyl ester, boc-d-serinemethyl ester, n-boc-d-serinemethylester, --methyl n-boc-d-serinate, r-methyl 2-tert-butoxycarbonyl amino-3-hydroxypropanoate, methyl n-tert-butoxycarbonyl-d-serinate
PubChem CID 377723
IUPAC Name methyl (2R)-3-hydroxy-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoate
SMILES CC(C)(C)OC(=O)NC(CO)C(=O)OC

Specifications

Boiling Point 215°C
Color Yellow
Quantity 5 g
Formula Weight 219.24
Percent Purity ≥98.0% (HPLC,N)
Physical Form Liquid
Chemical Name or Material N-(tert-Butoxycarbonyl)-D-serine Methyl Ester