missing translation for 'onlineSavingsMsg'
Learn More

N-(tert-Butoxycarbonyl)-beta-alanine 98.0+%, TCI America™

Catalog No. B21765G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B21765G 5 g
B217625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B21765G Supplier TCI America Supplier No. B21765G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 3303-84-2
Molecular Formula C8H15NO4
Molecular Weight (g/mol) 189.21
MDL Number MFCD00037291
InChI Key WCFJUSRQHZPVKY-UHFFFAOYSA-N
Synonym boc-beta-alanine, boc-beta-ala-oh, n-boc-beta-alanine, boc-b-ala-oh, boc-beta-ala, n-tert-butoxycarbonyl-beta-alanine, 3-tert-butoxycarbonyl amino propanoic acid, n-t-boc-beta-alanine, n-t-butyloxycarbonyl-beta-alanine, 3-tert-butoxycarbonylamino propanoic acid
PubChem CID 76809
IUPAC Name 3-{[(tert-butoxy)carbonyl]amino}propanoic acid
SMILES CC(C)(C)OC(=O)NCCC(O)=O

Specifications

Melting Point 77°C
Color White
Quantity 5 g
Formula Weight 189.21
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-(tert-Butoxycarbonyl)-beta-alanine