missing translation for 'onlineSavingsMsg'
Learn More

N-(tert-Butoxycarbonyl)-4-cyano-L-phenylalanine 98.0+%, TCI America™

Catalog No. B48775G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B48775G 5 g
B48771G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B48775G Supplier TCI America Supplier No. B48775G

Chemical Identifiers

CAS 131724-45-3
Molecular Formula C15H18N2O4
Molecular Weight (g/mol) 290.319
MDL Number MFCD00672526
InChI Key RMBLTLXJGNILPG-LBPRGKRZSA-N
Synonym boc-phe 4-cn-oh, boc-l-4-cyanophenylalanine, boc-4-cyano-l-phenylalanine, s-2-tert-butoxycarbonyl amino-3-4-cyanophenyl propanoic acid, boc-d-4-cyanophenylalanine, n-boc-4-cyanophenylalanine, s-n-boc-4-cyanophenylalanine, 2s-2-tert-butoxycarbonyl amino-3-4-cyanophenyl propanoic acid, 2s-3-4-cyanophenyl-2-2-methylpropan-2-yl oxycarbonylamino propanoic acid, boc-l-4-cn-phe-oh
PubChem CID 7020841
IUPAC Name (2S)-3-(4-cyanophenyl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
SMILES CC(C)(C)OC(=O)NC(CC1=CC=C(C=C1)C#N)C(=O)O

Specifications

Melting Point 147°C
Color White
Quantity 5 g
Formula Weight 290.32
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-(tert-Butoxycarbonyl)-4-cyano-L-phenylalanine