missing translation for 'onlineSavingsMsg'
Learn More

N-(tert-Butoxycarbonyl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline 99.0+%, TCI America™

Catalog No. B55885G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B55885G 5 g
B55881G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B55885G Supplier TCI America Supplier No. B55885G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 330793-01-6
Molecular Formula C17H26BNO4
Molecular Weight (g/mol) 319.21
MDL Number MFCD02179439
InChI Key HSJNIOYPTSKQBD-UHFFFAOYSA-N
Synonym 4-n-boc-amino phenylboronic acid pinacol ester, tert-butyl 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl carbamate, 4-boc-amino benzeneboronic acid pinacol ester, tert-butyl n-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl carbamate, 4-boc-amino phenylboronic acid pinacol cyclic ester, 4-boc-aminophenylboronic acid, pinacol ester, 4-tert-butoxycarbonyl aminophenylboronic acid, pinacol ester, tert-butyl n-4-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl carbamate, tert-butyl 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenylcarbamate
PubChem CID 2734617
IUPAC Name tert-butyl N-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]carbamate
SMILES CC(C)(C)OC(=O)NC1=CC=C(C=C1)B1OC(C)(C)C(C)(C)O1

Specifications

Melting Point 170°C
Color White
Quantity 5 g
Formula Weight 319.21
Percent Purity ≥99.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-(tert-Butoxycarbonyl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline