missing translation for 'onlineSavingsMsg'
Learn More

N-(tert-Butoxycarbonyl)-2-phosphonoglycine Trimethyl Ester 98.0+%, TCI America™

Catalog No. B40115G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B40115G 5 g
B40111G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B40115G Supplier TCI America Supplier No. B40115G
Only null left

Chemical Identifiers

CAS 89524-98-1
Molecular Formula C10H20NO7P
Molecular Weight (g/mol) 297.24
MDL Number MFCD02178893
InChI Key LJHAPRKTPAREGO-ZETCQYMHSA-N
Synonym methyl 2-tert-butoxycarbonyl amino-2-dimethoxyphosphoryl acetate, +/--boc-alpha-phosphonoglycine trimethyl ester, n-boc-2-phosphonoglycine trimethyl ester, methyl n-boc-2-dimethylphosphono glycinate, methyl 2-tert-butoxycarbonylamino-2-dimethoxyphosphoryl-acetate, boc-alpha-phosphonoglycine trimethyl ester, n-butoxycarbonyl trimethylphosphonoglycine, methyl 2-tert-butoxy carbonyl amino-2-dimethoxyphosphoryl acetate, boc-a-phosphonoglycine trimethyl ester, boc-?-phosphonoglycine trimethyl ester
PubChem CID 397380
IUPAC Name methyl (2S)-2-{[(tert-butoxy)carbonyl]amino}-2-(dimethoxyphosphoryl)acetate
SMILES COC(=O)[C@@H](NC(=O)OC(C)(C)C)P(=O)(OC)OC

Specifications

Melting Point 71°C
Color White
Quantity 5 g
Formula Weight 297.24
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material N-(tert-Butoxycarbonyl)-2-phosphonoglycine Trimethyl Ester