missing translation for 'onlineSavingsMsg'
Learn More

N-(tert-Butoxycarbonyl)-1,6-diaminohexane 98.0+%, TCI America™

Catalog No. A13755G
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A13755G 5 g
A13751G 1 g
A137525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A13755G Supplier TCI America Supplier No. A13755G

Chemical Identifiers

CAS 51857-17-1
Molecular Formula C11H24N2O2
Molecular Weight (g/mol) 216.325
MDL Number MFCD00671489
InChI Key RVZPDKXEHIRFPM-UHFFFAOYSA-N
Synonym n-boc-1,6-diaminohexane, n-boc-1,6-hexanediamine, tert-butyl n-6-aminohexyl carbamate, tert-butyl 6-aminohexyl carbamate, n-tert-butoxycarbonyl-1,6-diaminohexane, n-tert-butoxycarbonyl-1,6-hexanediamine, tert-butyl 6-aminohexylcarbamate, boc-1,6-diaminohexane hydrochloride, 6-tert-butoxycarbonylamino hexylamine, n-6-aminohexyl carbamic acid tert-butyl ester
PubChem CID 2733170
IUPAC Name tert-butyl N-(6-aminohexyl)carbamate
SMILES CC(C)(C)OC(=O)NCCCCCCN

Specifications

Quantity 5 g
Formula Weight 216.33
Percent Purity ≥97.0% (GC,T)
Physical Form Liquid
Chemical Name or Material N-(tert-Butoxycarbonyl)-1,6-diaminohexane
TSCA No
Recommended Storage Refrigerator