missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-(tert-Butoxycarbonyl)-1,3-diaminopropane 98.0+%, TCI America™
$144.30 - $457.57
Chemical Identifiers
| CAS | 75178-96-0 |
|---|---|
| Molecular Formula | C8H19N2O2 |
| Molecular Weight (g/mol) | 175.25 |
| MDL Number | MFCD00210021 |
| InChI Key | POHWAQLZBIMPRN-UHFFFAOYSA-O |
| Synonym | n-boc-1,3-propanediamine, n-boc-1,3-diaminopropane, tert-butyl n-3-aminopropyl carbamate, tert-butyl 3-aminopropyl carbamate, n-3-aminopropyl carbamic acid tert-butyl ester, tert-butyl 3-aminopropylcarbamate, 1-boc-amino-1,3-propanediamine, 3-amino-propyl-carbamic acid tert-butyl ester, n-t-boc-1,3-propanediamine, n-tert-butoxycarbonyl-1,3-diaminopropane |
| PubChem CID | 2735700 |
| IUPAC Name | tert-butyl N-(3-azaniumylpropyl)carbamate |
| SMILES | CC(C)(C)OC(=O)NCCC[NH3+] |
Chemical Identifiers
| 75178-96-0 | |
| 175.25 | |
| POHWAQLZBIMPRN-UHFFFAOYSA-O | |
| 2735700 | |
| CC(C)(C)OC(=O)NCCC[NH3+] |
| C8H19N2O2 | |
| MFCD00210021 | |
| n-boc-1,3-propanediamine, n-boc-1,3-diaminopropane, tert-butyl n-3-aminopropyl carbamate, tert-butyl 3-aminopropyl carbamate, n-3-aminopropyl carbamic acid tert-butyl ester, tert-butyl 3-aminopropylcarbamate, 1-boc-amino-1,3-propanediamine, 3-amino-propyl-carbamic acid tert-butyl ester, n-t-boc-1,3-propanediamine, n-tert-butoxycarbonyl-1,3-diaminopropane | |
| tert-butyl N-(3-azaniumylpropyl)carbamate |
Specifications
| 75178-96-0 | |
| White-Yellow | |
| MFCD00210021 | |
| n-boc-1,3-propanediamine, n-boc-1,3-diaminopropane, tert-butyl n-3-aminopropyl carbamate, tert-butyl 3-aminopropyl carbamate, n-3-aminopropyl carbamic acid tert-butyl ester, tert-butyl 3-aminopropylcarbamate, 1-boc-amino-1,3-propanediamine, 3-amino-propyl-carbamic acid tert-butyl ester, n-t-boc-1,3-propanediamine, n-tert-butoxycarbonyl-1,3-diaminopropane | |
| CC(C)(C)OC(=O)NCCC[NH3+] | |
| 175.25 | |
| 174.24 | |
| Crystal/Lump |
| 203°C | |
| C8H19N2O2 | |
| 3259 | |
| POHWAQLZBIMPRN-UHFFFAOYSA-O | |
| tert-butyl N-(3-azaniumylpropyl)carbamate | |
| 2735700 | |
| ≥98.0% (GC) | |
| N-(tert-Butoxycarbonyl)-1,3-diaminopropane |
Safety and Handling
TSCA : No