accessibility menu, dialog, popup

UserName

H3CCH3H3COOHNNH3

CAS RN 75178-96-0

IUPAC Name: tert-butyl N-(3-azaniumylpropyl)carbamate
Synonyms: tert-butyl N-(3-ammoniopropyl)carbamate
Molecular Weight (g/mol): 175.25
Molecular Formula: C8H19N2O2
InChi Key: POHWAQLZBIMPRN-UHFFFAOYSA-O
SMILES: CC(C)(C)OC(=O)NCCC[NH3+]
Percent Purity 
  • (2)
  • (2)
  • (1)
Quantity 
  • (2)
  • (2)
  • (1)
Boiling Point 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)