missing translation for 'onlineSavingsMsg'
Learn More

N-(tert-Butoxycarbonyl)-1,3-diaminopropane 98.0+%, TCI America™

Catalog No. A13721G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A13721G 1 g
A13725G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A13721G Supplier TCI America Supplier No. A13721G

Chemical Identifiers

CAS 75178-96-0
Molecular Formula C8H19N2O2
Molecular Weight (g/mol) 175.25
MDL Number MFCD00210021
InChI Key POHWAQLZBIMPRN-UHFFFAOYSA-O
Synonym n-boc-1,3-propanediamine, n-boc-1,3-diaminopropane, tert-butyl n-3-aminopropyl carbamate, tert-butyl 3-aminopropyl carbamate, n-3-aminopropyl carbamic acid tert-butyl ester, tert-butyl 3-aminopropylcarbamate, 1-boc-amino-1,3-propanediamine, 3-amino-propyl-carbamic acid tert-butyl ester, n-t-boc-1,3-propanediamine, n-tert-butoxycarbonyl-1,3-diaminopropane
PubChem CID 2735700
IUPAC Name tert-butyl N-(3-azaniumylpropyl)carbamate
SMILES CC(C)(C)OC(=O)NCCC[NH3+]

Specifications

Boiling Point 203°C
Color White-Yellow
UN Number 3259
Quantity 1 g
Formula Weight 174.24
Percent Purity ≥98.0% (GC)
Physical Form Crystal/Lump
Chemical Name or Material N-(tert-Butoxycarbonyl)-1,3-diaminopropane