missing translation for 'onlineSavingsMsg'
Learn More

Nω-(2-Acetamido-3,4,6-tri-O-benzyl-2-deoxy-β-D-glucopyranosyl)-Nα-Boc-L-asparagine Benzyl Ester 97+%, TCI America™

Catalog No. A1685100MG
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A1685100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A1685100MG Supplier TCI America Supplier No. A1685100MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 219968-28-2
Molecular Formula C45H53N3O10
Molecular Weight (g/mol) 795.93
MDL Number MFCD06797050
InChI Key CRNGSGNVFXQOBD-UHFFFAOYNA-N
PubChem CID 15458580
IUPAC Name benzyl 3-{[4,5-bis(benzyloxy)-6-[(benzyloxy)methyl]-3-acetamidooxan-2-yl]carbamoyl}-2-{[(tert-butoxy)carbonyl]amino}propanoate
SMILES CC(=O)NC1C(NC(=O)CC(NC(=O)OC(C)(C)C)C(=O)OCC2=CC=CC=C2)OC(COCC2=CC=CC=C2)C(OCC2=CC=CC=C2)C1OCC1=CC=CC=C1

Specifications

Melting Point 214°C
Color White-Yellow
Quantity 100 mg
Formula Weight 795.93
Percent Purity ≥97.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Nomega-(2-Acetamido-3,4,6-tri-O-benzyl-2-deoxy-beta-D-glucopyranosyl)-Nalpha-(tert-butoxycarbonyl)-L
TSCA No
Recommended Storage Freezer