missing translation for 'onlineSavingsMsg'
Learn More

N-Succinimidyl 4-Maleimidobutyrate 98,0+%, TCI America™

Numéro de catalogue. S0399100MG
Modifier l'affichage
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
S0399100MG 100 mg
S03991G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. S0399100MG Fournisseur TCI America Code fournisseur S0399100MG

Identifiants chimiques

CAS 80307-12-6
Molecular Formula C12H12N2O6
Molecular Weight (g/mol) 280.24
MDL Number MFCD00036817
InChI Key PVGATNRYUYNBHO-UHFFFAOYSA-N
Synonym gmbs, 4-maleimidobutyric acid n-hydroxysuccinimide ester, 4-maleimidobutyric acid n-succinimidyl ester, n-gamma-maleimidobutyryloxy succinimide, 2,5-dioxopyrrolidin-1-yl 4-2,5-dioxo-2,5-dihydro-1h-pyrrol-1-yl butanoate, n-4-maleimidobutyryloxy succinimide, n-succinimidyl 4-maleimidobutyrate, n-succinimidyl-4-maleimidobutyrate, 2,5-dioxopyrrolidin-1-yl 4-2,5-dioxopyrrol-1-yl butanoate
PubChem CID 133440
IUPAC Name 2,5-dioxopyrrolidine-1-yl 4-(2,5-dioxo-2,5-dihydro-1H-pyrrol-1-yl) butanoate
SMILES O=C(CCCN1C(=O)C=CC1=O)ON1C(=O)CCC1=O

Spécifications

Melting Point 130°C
Quantity 100 mg
Formula Weight 280.24
Percent Purity ≥98.0% (HPLC,N)
Physical Form Poudre cristalline
Chemical Name or Material N-Succinimidyl 4-Maleimidobutyrate [Cross-linking Reagent]
TSCA No
Recommended Storage Congélateur (-20 °C)