missing translation for 'onlineSavingsMsg'
Learn More

N-Succinimidyl 4-[4-(Dimethylamino)phenylazo]benzoate 98.0+%, TCI America™

Catalog No. S0857200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S0857200MG 200 mg
S08571G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S0857200MG Supplier TCI America Supplier No. S0857200MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 146998-31-4
Molecular Formula C19H18N4O4
Molecular Weight (g/mol) 366.377
MDL Number MFCD00467415
InChI Key IBOVDNBDQHYNJI-UHFFFAOYSA-N
Synonym 2,5-dioxopyrrolidin-1-yl 4-4-dimethylamino phenyl diazenyl benzoate, dabcyl se dye, dabcyl acid, se, 4-4-dimethylamino phenyl azo benzoic acid, succinimidyl ester, 4-4-dimethylamino phenylazo benzoic acid n-succinimidyl ester, dabcyl-nhs, e-2,5-dioxopyrrolidin-1-yl 4-4-dimethylamino phenyl diazenyl benzoate, 2,5-pyrrolidinedione, 1-4-4-dimethylamino phenyl azo benzoyl oxy, dabcyl-se; 4-4-dimethylamino phenylazo benzoic acid n-succinimidyl ester, dabcyl se, solid
PubChem CID 4206604
ChEBI CHEBI:52005
IUPAC Name (2,5-dioxopyrrolidin-1-yl) 4-[[4-(dimethylamino)phenyl]diazenyl]benzoate
SMILES CN(C)C1=CC=C(C=C1)N=NC2=CC=C(C=C2)C(=O)ON3C(=O)CCC3=O

Specifications

Melting Point 242°C
Quantity 200 mg
Formula Weight 366.38
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-Succinimidyl 4-[4-(Dimethylamino)phenylazo]benzoate
TSCA No
Recommended Storage Refrigerator