missing translation for 'onlineSavingsMsg'
Learn More

N-(Pyrazinylcarbonyl)-L-phenylalanine 98.0+%, TCI America™

Catalog No. P20681G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P20681G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P20681G Supplier TCI America Supplier No. P20681G

Chemical Identifiers

CAS 114457-94-2
Molecular Formula C14H13N3O3
Molecular Weight (g/mol) 271.276
MDL Number MFCD09800536
InChI Key DWYZPDHMMZGQAP-NSHDSACASA-N
PubChem CID 15042559
IUPAC Name (2S)-3-phenyl-2-(pyrazine-2-carbonylamino)propanoic acid
SMILES C1=CC=C(C=C1)CC(C(=O)O)NC(=O)C2=NC=CN=C2

Specifications

Melting Point 169°C
Color White
Quantity 1 g
Formula Weight 271.28
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material N-(Pyrazinylcarbonyl)-L-phenylalanine