missing translation for 'onlineSavingsMsg'
Learn More

N-Propyl-N-(2,3-epoxypropyl)perfluoro-n-octylsulfonamide 80.0+%, TCI America™

Catalog No. P116325G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P116325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P116325G Supplier TCI America Supplier No. P116325G
Only null left

Chemical Identifiers

CAS 77620-64-5
Molecular Formula C14H12F17NO3S
Molecular Weight (g/mol) 597.29
MDL Number MFCD00191486
InChI Key HEAWIZXIWLKJKG-UHFFFAOYNA-N
PubChem CID 4712723
IUPAC Name 1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-heptadecafluoro-N-[(oxiran-2-yl)methyl]-N-propyloctane-1-sulfonamide
SMILES CCCN(CC1CO1)S(=O)(=O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F

Specifications

Color Yellow
Quantity 25 g
Formula Weight 597.29
Percent Purity ≥80.0% (GC)
Physical Form Liquid
Chemical Name or Material N-Propyl-N-(2,3-epoxypropyl)perfluoro-n-octylsulfonamide