missing translation for 'onlineSavingsMsg'
Learn More

n-Propyl Alcohol, for HPLC, GC and spectrophotometry, >99.5%, Solstice

Catalog No. 6000103
Change view
Click to view available options
Quantity:
4 x 4 L
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
6000103 4 x 4 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 6000103 Supplier Solstice Supplier No. 3224

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 71-23-8
Molecular Formula C3H8O
Molecular Weight (g/mol) 60.10
MDL Number MFCD00002941
InChI Key BDERNNFJNOPAEC-UHFFFAOYSA-N
Synonym 1-propanol, propanol, propyl alcohol, n-propanol, n-propyl alcohol, ethylcarbinol, 1-hydroxypropane, optal, osmosol extra, propylic alcohol
PubChem CID 1031
ChEBI CHEBI:28831
IUPAC Name propan-1-ol
SMILES CCCO

Specifications

Absorbance 0.5 AU max. at 225 nm, 0.05 AU max. at 250 nm, 0.005 AU max. at 300 nm, 0.005 AU max. at 400 nm
CAS Max % 100.0000%
Density 0.8053 g/cm3
Quantity 4 x 4 L
Flash Point 23°C
Packaging Glass Bottle
UN Number UN1274
Beilstein 1098242
Boiling Point 97.2°C
Formula Weight 60.09
Percent Purity >99.5%
Chemical Name or Material n-Propyl Alcohol, B&J Brand™, for HPLC, GC and spectrophotometry, >99.5%
Show More Show Less
ShelfLife 547 days from date of manufacture
DOTInformation Transport Hazard Class: 3; Packing Group:3: III; Proper Shipping Name: n-Propanol