missing translation for 'onlineSavingsMsg'
Learn More

N-Propyl Acetate, Reagent Grade, Reagents

Catalog No. 01338133
Change view
Click to view available options
Quantity:
4 L
10 L
20 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01338133 10 L
01338132 20 L
01338131 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01338133 Supplier Labchem Inc Supplier No. C112101010E6

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

N-Propyl Acetate, Reagent Grade, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 109-60-4
Molecular Formula C5H10O2
Molecular Weight (g/mol) 102.13
InChI Key YKYONYBAUNKHLG-UHFFFAOYSA-N
Synonym Propyl Ethanoate, Acetic Acid Propyl Ester
IUPAC Name propyl acetate
SMILES CCCOC(C)=O

Specifications

Melting Point -93°C
Color Colorless
Boiling Point 101.5°C
Packaging Steel Pail
Quantity 10 L
Specific Gravity 0.886
Grade Reagent
Physical Form Liquid
Chemical Name or Material N-Propyl Acetate, Reagent Grade