missing translation for 'onlineSavingsMsg'
Learn More

N-Phthaloyl-L-phenylalanine 98.0+%, TCI America™

Numéro de catalogue. P12275G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
P12275G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. P12275G Fournisseur TCI America Code fournisseur P12275G
Il en reste null

Identifiants chimiques

CAS 5123-55-7
Molecular Formula C17H13NO4
Molecular Weight (g/mol) 295.29
MDL Number MFCD00069738
InChI Key VAYRSTHMTWUHGE-AWEZNQCLSA-N
Synonym n-phthaloyl-l-phenylalanine, n-pht-phe-oh, phthaloyl-l-phenylalanine, n-phthalyl-l-phenylalanine, s-n-phthaloylphenylalanine, n-phthaloyl-phe-oh, 2s-2-1,3-dioxoisoindol-2-yl-3-phenylpropanoic acid, 2s-2-1,3-dioxo-2,3-dihydro-1h-isoindol-2-yl-3-phenylpropanoic acid, pht-phe-oh, ncistruc1_000419
PubChem CID 334207
IUPAC Name (2S)-2-(1,3-dioxo-2,3-dihydro-1H-isoindol-2-yl)-3-phenylpropanoic acid
SMILES OC(=O)[C@H](CC1=CC=CC=C1)N1C(=O)C2=CC=CC=C2C1=O

Spécifications

Melting Point 185°C
Color White
Quantity 5 g
Formula Weight 295.29
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-Phthaloyl-L-phenylalanine