missing translation for 'onlineSavingsMsg'
Learn More

N''-Phenylguanidine nitrate, ≥97%, Thermo Scientific™

Catalog No. BTB10165DA
Change view
Click to view available options
Quantity:
1g
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
BTB10165DA 1g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BTB10165DA Supplier Thermo Scientific Chemicals Supplier No. BTB10165DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 18860-78-1
Molecular Formula C7H10N4O3
Molecular Weight (g/mol) 198.182
MDL Number MFCD00213415
InChI Key DIHKZQUSGDYVOT-UHFFFAOYSA-N
Synonym phenylguanidine nitrate, 1-phenylguanidine nitrate, 2-phenylguanidine nitrate, n-phenylguanidine nitrate, 2-phenylguanidine; nitric acid, nitric acid; 2-phenylguanidine, nitric acid compound with n-phenylguanidine 1:1, phenyl guanidine nitrate
PubChem CID 2775226
IUPAC Name nitric acid;2-phenylguanidine
SMILES C1=CC=C(C=C1)N=C(N)N.[N+](=O)(O)[O-]

Specifications

Assay Percent Range 97+%
Packaging Amber Glass Bottle
Quantity 1g
Formula Weight 198.18
Percent Purity 97%
Chemical Name or Material N''-phenylguanidine nitrate
Hazard Statement GHS H Statement:
Harmful in contact with skin.
Harmful if swallowed.
Causes serious eye irritation.
Harmful if inhaled.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.