missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Phenylglycine 97.0+%, TCI America™
Spécifications
Spécifications
| Melting Point | 125°C |
| Color | White-Yellow |
| Quantity | 25 g |
| Formula Weight | 151.17 |
| Percent Purity | ≥97.0% (T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | N-Phenylglycine |
Sécurité et manipulation
| EINECSNumber | (3)-1441 |
| TSCA | Yes |