missing translation for 'onlineSavingsMsg'
Learn More

N-Phenylbis(trifluoromethanesulfonimide), 99%

Catalog No. AAL1253106
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1253106 5 g
AAL1253114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1253106 Supplier Thermo Scientific Chemicals Supplier No. L1253106
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 37595-74-7 | C8H5F6NO4S2 | 357.241 g/mol

N-Phenylbis(trifluoromethanesulfonimide) acts as a mild triflating reagent as well as a transparent strong electron-withdrawing p-type dopant in carbon nanotubes. It is also employed as a reactant for the preparation of amphoteric alfa-boryl aldehydes. It plays an important role in the enantioselective synthesis of the core ring skeleton of leucosceptroids A-D and in steroselective sulfoxidation. Further, it is used in the enantioselective synthesis of beta-amino acids through the Mannich reaction. In addition to this, it is used in the preparation of sphingosine 1-phosphate-1 receptor agonists.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Phenylbis(trifluoromethanesulfonimide) acts as a mild triflating reagent as well as a transparent strong electron-withdrawing p-type dopant in carbon nanotubes. It is also employed as a reactant for the preparation of amphoteric alfa-boryl aldehydes. It plays an important role in the enantioselective synthesis of the core ring skeleton of leucosceptroids A-D and in steroselective sulfoxidation. Further, it is used in the enantioselective synthesis of beta-amino acids through the Mannich reaction. In addition to this, it is used in the preparation of sphingosine 1-phosphate-1 receptor agonists.

Solubility
Soluble in methanol. Slightly soluble in chloroform and ethyl acetate.

Notes
Hygroscopic. Store in a cool place. Moisture sensitive. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 37595-74-7
Molecular Formula C8H5F6NO4S2
Molecular Weight (g/mol) 357.241
MDL Number MFCD00000404
InChI Key DIOHEXPTUTVCNX-UHFFFAOYSA-N
Synonym n,n-bis trifluoromethylsulfonyl aniline, phenyl triflimide, n-phenyl-bis trifluoromethanesulfonimide, n-phenyltrifluoromethanesulfonimide, n,n-bis-trifluoromethanesulfonyl aniline, 1,1,1-trifluoro-n-phenyl-n-trifluoromethylsulfonyl methanesulfonamide, n-phenylbis trifluoromethanesulfonimide, n-phenylbis trifluoromethanesulphonimide, 1,1,1-trifluoro-n-phenyl-n-trifluoromethyl sulfonyl methanesulfonamide, n,n-bis trifluoromethanesulfonyl aniline
PubChem CID 142176
IUPAC Name 1,1,1-trifluoro-N-phenyl-N-(trifluoromethylsulfonyl)methanesulfonamide
SMILES C1=CC=C(C=C1)N(S(=O)(=O)C(F)(F)F)S(=O)(=O)C(F)(F)F

Specifications

Melting Point 100°C to 102°C
Quantity 5 g
Beilstein 1269141
Sensitivity Moisture sensitive
Solubility Information Soluble in methanol. Slightly soluble in chloroform and ethyl acetate.
Formula Weight 357.25
Percent Purity 99%
Chemical Name or Material N-Phenylbis(trifluoromethanesulfonimide)
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only