Learn More
N-Phenylanthranilic acid, 99%
CAS: 91-40-7 | C13H11NO2 | 213.236 g/mol
Supplier: Thermo Scientific Chemicals A1434022
Description
DPC is a potent, non-specific blocker of Cl- channel and chloride-bicarbonate exchange. DPC also inhibits CFTR (cystic fibrosis transmembrane regulator) ATPase activity. It is used to detect vanadium in steel.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsDPC is a potent, non-specific blocker of Cl- channel and chloride-bicarbonate exchange. DPC also inhibits CFTR (cystic fibrosis transmembrane regulator) ATPase activity. It is used to detect vanadium in steel.
Solubility
Soluble in ethanol, DMSO (50 mg/ml), hot alcohol, and methanol. Insoluble in water.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications
| N-Phenylanthranilic acid | |
| 185°C to 188°C | |
| MFCD00002421 | |
| 1456607 | |
| n-phenylanthranilic acid, 2-phenylamino benzoic acid, fenamic acid, diphenylamine-2-carboxylic acid, phenylanthranilic acid, 2-carboxydiphenylamine, benzoic acid, 2-phenylamino, o-anilinobenzoic acid, n-phenyl-o-aminobenzoic acid, n-phenyl-2-aminobenzoic acid | |
| ZWJINEZUASEZBH-UHFFFAOYSA-N | |
| 2-anilinobenzoic acid | |
| 4386 | |
| 213.24 |
| 91-40-7 | |
| C13H11NO2 | |
| 100 g | |
| 14,7273 | |
| Soluble in ethanol,DMSO (50mg/ml),hot alcohol,and methanol. Insoluble in water. | |
| C1=CC=C(C=C1)NC2=CC=CC=C2C(=O)O | |
| 213.236 | |
| CHEBI:34756 | |
| 99% |
Chemical Identifiers
| 91-40-7 | |
| 213.236 | |
| ZWJINEZUASEZBH-UHFFFAOYSA-N | |
| 4386 | |
| 2-anilinobenzoic acid |
| C13H11NO2 | |
| MFCD00002421 | |
| n-phenylanthranilic acid, 2-phenylamino benzoic acid, fenamic acid, diphenylamine-2-carboxylic acid, phenylanthranilic acid, 2-carboxydiphenylamine, benzoic acid, 2-phenylamino, o-anilinobenzoic acid, n-phenyl-o-aminobenzoic acid, n-phenyl-2-aminobenzoic acid | |
| CHEBI:34756 | |
| C1=CC=C(C=C1)NC2=CC=CC=C2C(=O)O |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 202-066-8
RTECSNumber : CB3730000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only