CAS RN 91-40-7
IUPAC Name:
2-(phenylamino)benzoic acid
Synonyms:
phenylanthranilic acid 2-carboxydiphenylamine fenamic acid dpc cpd fenamic acid, ca salt (2:1) monosodium salt fenamic acid N-phenyl-o-aminobenzoic acid ca salt (2:1) fenamic acid diphenylamine-2-carboxylate fenamic acid, monosodium salt
Molecular Weight (g/mol):
213.24
Molecular Formula:
C13H11NO2
InChi Key:
ZWJINEZUASEZBH-UHFFFAOYSA-N
SMILES:
OC(=O)C1=CC=CC=C1NC1=CC=CC=C1
1
–
1
of
1
results
1
Promotions Available
N-Phenylanthranilic acid, 99%
N-Phenylanthranilic acid, 99%
CAS: 91-40-7 Molecular Formula: C13H11NO2 Molecular Weight (g/mol): 213.236 MDL Number: MFCD00002421 InChI Key: ZWJINEZUASEZBH-UHFFFAOYSA-N Synonym: n-phenylanthranilic acid,2-phenylamino benzoic acid,fenamic acid,diphenylamine-2-carboxylic acid,phenylanthranilic acid,2-carboxydiphenylamine,benzoic acid, 2-phenylamino,o-anilinobenzoic acid,n-phenyl-o-aminobenzoic acid,n-phenyl-2-aminobenzoic acid PubChem CID: 4386 ChEBI: CHEBI:34756 IUPAC Name: 2-anilinobenzoic acid SMILES: C1=CC=C(C=C1)NC2=CC=CC=C2C(=O)O