missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Phenylanthranilic acid, 99%
Description
DPC is a potent, non-specific blocker of Cl- channel and chloride-bicarbonate exchange. DPC also inhibits CFTR (cystic fibrosis transmembrane regulator) ATPase activity. It is used to detect vanadium in steel.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
DPC is a potent, non-specific blocker of Cl- channel and chloride-bicarbonate exchange. DPC also inhibits CFTR (cystic fibrosis transmembrane regulator) ATPase activity. It is used to detect vanadium in steel.
Solubility
Soluble in ethanol, DMSO (50 mg/ml), hot alcohol, and methanol. Insoluble in water.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
DPC is a potent, non-specific blocker of Cl- channel and chloride-bicarbonate exchange. DPC also inhibits CFTR (cystic fibrosis transmembrane regulator) ATPase activity. It is used to detect vanadium in steel.
Solubility
Soluble in ethanol, DMSO (50 mg/ml), hot alcohol, and methanol. Insoluble in water.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications
Specifications
| Melting Point | 185°C to 188°C |
| Quantity | 25 g |
| Beilstein | 1456607 |
| Merck Index | 14,7273 |
| Solubility Information | Soluble in ethanol,DMSO (50mg/ml),hot alcohol,and methanol. Insoluble in water. |
| Formula Weight | 213.24 |
| Percent Purity | 99% |
| Chemical Name or Material | N-Phenylanthranilic acid |
Safety and Handling
| Hazard Category | H315-H319-H335 |
| Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
| EINECSNumber | 202-066-8 |
| RTECSNumber | CB3730000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only