missing translation for 'onlineSavingsMsg'
Learn More

N-Phenyl-o-phenylenediamine, 98%

Catalog No. AAL1161606
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1161606 5 g
AAL1161614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1161606 Supplier Thermo Scientific Chemicals Supplier No. L1161606
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 534-85-0 | C12H12N2 | 184.24 g/mol

N-Phenyl-o-phenylenediamine is also used as an intermediate in organic synthesis and pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Phenyl-o-phenylenediamine is also used as an intermediate in organic synthesis and pharmaceuticals.

Solubility
Soluble in acetone, chloroform and benzene. Insoluble in water.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers. It is sensitive to light. Store at room temperature. Incompatible with acid chlorides, acid anhydrides, chloroformates and strong oxidizing agents.

Chemical Identifiers

CAS 534-85-0
Molecular Formula C12H12N2
Molecular Weight (g/mol) 184.24
MDL Number MFCD00007685
InChI Key NFCPRRWCTNLGSN-UHFFFAOYSA-N
Synonym 2-aminodiphenylamine, n-phenyl-o-phenylenediamine, n1-phenylbenzene-1,2-diamine, o-semidine, 1,2-benzenediamine, n-phenyl, o-aminodiphenylamine, n-phenyl-1,2-phenylenediamine, n-phenylbenzene-1,2-diamine, o-phenylenediamine, n-phenyl, unii-g2x60k1y26
PubChem CID 68297
IUPAC Name 2-N-phenylbenzene-1,2-diamine
SMILES NC1=CC=CC=C1NC1=CC=CC=C1

Specifications

Melting Point 77°C to 80°C
Quantity 5 g
Beilstein 908984
Solubility Information Soluble in acetone,chloroform and benzene. Insoluble in water.
Formula Weight 184.24
Percent Purity 98%
Chemical Name or Material N-Phenyl-o-phenylenediamine
Hazard Category H302-H312-H315-H319-H332-H335-H341
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P201-P202-P261-P264b-P270-P271-P280i-P281-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P312-P330-P332+P313-P362-P501c
EINECSNumber 208-606-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only