missing translation for 'onlineSavingsMsg'
Learn More

N-Phenyl-3-biphenylamine 98.0+%, TCI America™

Catalog No. P1497500MG
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P1497500MG 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P1497500MG Supplier TCI America Supplier No. P1497500MG

Chemical Identifiers

CAS 198275-79-5
Molecular Formula C18H15N
Molecular Weight (g/mol) 245.33
MDL Number MFCD08276409
InChI Key QJAYWJUCAONYLG-UHFFFAOYSA-N
Synonym 3-Anilinobiphenyl
PubChem CID 20615412
IUPAC Name N-phenyl-[1,1'-biphenyl]-3-amine
SMILES N(C1=CC=CC=C1)C1=CC=CC(=C1)C1=CC=CC=C1

Specifications

Melting Point 92°C
Color Green-Yellow
Quantity 500 mg
Formula Weight 245.33
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material N-Phenyl-3-biphenylamine